Learn More
Medchemexpress LLC HY-15102 5mg Medchemexpress, MK-0429 CAS:227963-15-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY151025MG
Encompass_Preferred
Medchemexpress, HY-15102 5mg MK-0429 CAS:227963-15-7 MK-0429 (L-000845704) is an orally active, potent, selective and nonpeptide αvβ3 integrin antagonist with an IC 50 of 80 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2724