Learn More
Medchemexpress LLC Mk-0493 | 455956-93-1 | 99.4% | 546.09 | C30H38ClF2N3O2 | 5 MG
SDP
Catalog No. 5000228082
Encompass_Preferred
MK-0493 is a potent, orally active, and selective agonist of the melanocortin receptor 4 (MC4R) used in research to investigate appetite regulation and energy homeostasis. The compound is supplied as a powder with high analytical purity suitable for formulation and biological testing.
  • Potent, selective MC4R agonist
  • Orally active small molecule
  • Demonstrated reduction in energy intake in preclinical studies
  • High purity (99.4% by RP-HPLC)
  • Provided as a powder for solution preparation
Catalog No. 50-002-28082 Supplier Medchemexpress LLC Supplier No. HY1189305MG
MK-0493 is a potent, orally active, and selective agonist of the melanocortin receptor 4 (MC4R) used in research to investigate appetite regulation and energy homeostasis. The compound is supplied as a powder with high analytical purity suitable for formulation and biological testing.
  • Potent, selective MC4R agonist
  • Orally active small molecule
  • Demonstrated reduction in energy intake in preclinical studies
  • High purity (99.4% by RP-HPLC)
  • Provided as a powder for solution preparation