Learn More
Medchemexpress LLC MK-0493 hydrochloride | 455957-71-8 | 98.8% | 582.55 g/mol | C30H39Cl2F2N3O2 | 5 MG
SDP
Catalog No. 5000307910
Encompass_Preferred
MK-0493 hydrochloride is a potent, orally active, and selective agonist of the melanocortin receptor 4 (MC4R) used as a research tool to study appetite regulation and energy homeostasis. It is supplied as a high-purity hydrochloride salt suitable for analytical and preclinical applications.
  • Potent, selective MC4R agonist for receptor pharmacology studies.
  • Orally active probe for appetite and energy intake research.
  • High purity (98.8%) solid suitable for analytical and preclinical use.
  • Available in mg-scale solid quantities and as a DMSO solution.
  • Soluble in DMSO; handle and store according to standard compound precautions.
Catalog No. 50-003-07910 Supplier Medchemexpress LLC Supplier No. HY118930A5MG
MK-0493 hydrochloride is a potent, orally active, and selective agonist of the melanocortin receptor 4 (MC4R) used as a research tool to study appetite regulation and energy homeostasis. It is supplied as a high-purity hydrochloride salt suitable for analytical and preclinical applications.
  • Potent, selective MC4R agonist for receptor pharmacology studies.
  • Orally active probe for appetite and energy intake research.
  • High purity (98.8%) solid suitable for analytical and preclinical use.
  • Available in mg-scale solid quantities and as a DMSO solution.
  • Soluble in DMSO; handle and store according to standard compound precautions.