Learn More
Medchemexpress LLC HY-15411 200mg Medchemexpress, MK-0557 CAS:328232-95-7 Purity:98%
SDP
Catalog No. 501638315
Encompass_Preferred
Medchemexpress,HY-15411 200mg MK-0557 CAS:328232-95-7 Formula:C22H19FN4O3 MK-0557 is a highly selective, orally available neuropeptide Y5 receptor antagonist with a Ki of 1.6 nM. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-8315 Supplier Medchemexpress LLC Supplier No. HY15411200MG
Medchemexpress,HY-15411 200mg MK-0557 CAS:328232-95-7 Formula:C22H19FN4O3 MK-0557 is a highly selective, orally available neuropeptide Y5 receptor antagonist with a Ki of 1.6 nM. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.