Learn More
Medchemexpress LLC MK-0752 | 471905-41-6 | 99.35% | 442.90 | 100 UG
SDP
Catalog No. 5000344447
Encompass_Preferred
MK-0752 is a potent, orally active, and specific γ-secretase inhibitor. It demonstrates a dose-dependent reduction of Aβ40 with an IC50 of 5 nM in human SH-SY5Y cells. MK-0752 is capable of crossing the blood-brain barrier and reduces newly generated CNS Aβ in vivo. It is for research use only.
  • Potent, orally active, and specific γ-secretase inhibitor.
  • Dose-dependent reduction of Aβ40 with an IC50 of 5 nM in human SH-SY5Y cells.
  • Crosses the blood-brain barrier.
  • Reduces newly generated CNS Aβ in vivo.
Catalog No. 50-003-44447 Supplier Medchemexpress LLC Supplier No. HY109741MG
MK-0752 is a potent, orally active, and specific γ-secretase inhibitor. It demonstrates a dose-dependent reduction of Aβ40 with an IC50 of 5 nM in human SH-SY5Y cells. MK-0752 is capable of crossing the blood-brain barrier and reduces newly generated CNS Aβ in vivo. It is for research use only.
  • Potent, orally active, and specific γ-secretase inhibitor.
  • Dose-dependent reduction of Aβ40 with an IC50 of 5 nM in human SH-SY5Y cells.
  • Crosses the blood-brain barrier.
  • Reduces newly generated CNS Aβ in vivo.