Learn More
Medchemexpress LLC HY-11027 5mg Medchemexpress, MK-0773 CAS:606101-58-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY110275MG
Encompass_Preferred
Medchemexpress, HY-11027 5mg MK-0773 CAS:606101-58-0 MK-0773 is a selective androgen receptor modulators (SARMs) that binds to AR with an IC50 of 6.6 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8785