Learn More
Medchemexpress LLC MK-0812 (succinate) | 851916-42-2 | 99.9% | C28H40F3N3O7 | 5 MG
SDP
Catalog No. 5000359113
Encompass_Preferred
MK-0812 succinate is a potent and selective CCR2 antagonist with high affinity. It completely blocks all MCP-1 mediated responses in a concentration-dependent manner. This compound is intended for research use only.
  • Potent and selective CCR2 antagonist with high affinity
  • Completely blocks all MCP-1 mediated responses (IC50 3.2 nM)
  • Inhibits 125I-MCP-1 binding on isolated monocytes (IC50 4.5 nM)
  • Inhibits MCP-1 induced monocyte shape change in whole blood assays (IC50 8 nM)
  • Reduces Ly6G-Ly6Chi monocytes in peripheral blood in vivo
  • Causes dose-dependent reduction in circulating Ly6Chi monocytes in vivo
Catalog No. 50-003-59113 Supplier Medchemexpress LLC Supplier No. HY50669A5MG
MK-0812 succinate is a potent and selective CCR2 antagonist with high affinity. It completely blocks all MCP-1 mediated responses in a concentration-dependent manner. This compound is intended for research use only.
  • Potent and selective CCR2 antagonist with high affinity
  • Completely blocks all MCP-1 mediated responses (IC50 3.2 nM)
  • Inhibits 125I-MCP-1 binding on isolated monocytes (IC50 4.5 nM)
  • Inhibits MCP-1 induced monocyte shape change in whole blood assays (IC50 8 nM)
  • Reduces Ly6G-Ly6Chi monocytes in peripheral blood in vivo
  • Causes dose-dependent reduction in circulating Ly6Chi monocytes in vivo