Learn More
TARGETMOL CHEMICALS INC MK-0873 5MG
SDP
Catalog No. 502580308
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. MK-0873 is a novel and potent selective phosphodiesterase 4 (PDE4) inhibitor. purity: 99%
Catalog No. 50-258-0308 Supplier TARGETMOL CHEMICALS INC Supplier No. T334225MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. MK-0873 is a novel and potent selective phosphodiesterase 4 (PDE4) inhibitor. purity: 99%