Learn More
Apexbio Technology LLC MK 0893 870823-12-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063005
Encompass_Preferred
MK 0893 is a small molecule antagonist targeting both the glucagon receptor (GCGR) and insulin-like growth factor-1 receptor (IGF-1R) It competitively and reversibly inhibits glucagon binding to GCGR subsequently reducing receptor-mediated cAMP production In cell-based assays utilizing CHO cells expressing human GCGR MK 0893 showed nanomolar-level inhibitory potency with a receptor binding IC50 around 6 6 nM and a cellular cAMP inhibition IC50 near 15 7 nM Additionally MK 0893 exhibits potent inhibitory activity on IGF-1R demonstrated in IGF-1-driven mouse xenograft tumor models Therefore MK 0893 serves as a relevant pharmacological tool in research related to type 2 diabetes and IGF-1R-mediated oncogenic signaling pathways
Catalog No. 50-000-63005 Supplier Apexbio Technology LLC Supplier No. A36085.1
MK 0893 is a small molecule antagonist targeting both the glucagon receptor (GCGR) and insulin-like growth factor-1 receptor (IGF-1R) It competitively and reversibly inhibits glucagon binding to GCGR subsequently reducing receptor-mediated cAMP production In cell-based assays utilizing CHO cells expressing human GCGR MK 0893 showed nanomolar-level inhibitory potency with a receptor binding IC50 around 6 6 nM and a cellular cAMP inhibition IC50 near 15 7 nM Additionally MK 0893 exhibits potent inhibitory activity on IGF-1R demonstrated in IGF-1-driven mouse xenograft tumor models Therefore MK 0893 serves as a relevant pharmacological tool in research related to type 2 diabetes and IGF-1R-mediated oncogenic signaling pathways