Learn More
Medchemexpress LLC HY-19843 5mg Medchemexpress, MK-0941 CAS:1137916-97-2 Purity:>98%
SDP
Catalog No. 501970039
Encompass_Preferred
Medchemexpress, HY-19843 5mg MK-0941 CAS:1137916-97-2 MK-0941 is a potent, orally active and allosteric glucokinase activator, with EC50s of 240 and 65 nM for recombinant human glucokinase in the presence of 2.5 and 10 mM glucose, respectively. MK-0941 has potential in the treatment of type 2 diabetes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0039 Supplier Medchemexpress LLC Supplier No. HY198435MG
Medchemexpress, HY-19843 5mg MK-0941 CAS:1137916-97-2 MK-0941 is a potent, orally active and allosteric glucokinase activator, with EC50s of 240 and 65 nM for recombinant human glucokinase in the presence of 2.5 and 10 mM glucose, respectively. MK-0941 has potential in the treatment of type 2 diabetes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.