Learn More
Selleck Chemical LLC Adavosertib (MK-1775) 10mM/1mL 955365-80-7 AZD1775
SDP
Supplier:  Selleck Chemical LLC S152510MM/1ML
Encompass_Preferred
Adavosertib (MK-1775, AZD1775) is a potent and selective Wee1 inhibitor with IC50 of 5.2 nM in a cell-free assay; hinders G2 DNA damage checkpoint. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1097015