Learn More
Medchemexpress LLC HY-10993 100mg , MK-1775 AZD1775;Adavosertib CAS:955365-80-7 Purity:98%
SDP
Catalog No. 501492669
Encompass_Preferred
Medchemexpress, HY-10993 100mg MK-1775 AZD1775;Adavosertib CAS:955365-80-7 Formula:C27H32N8O2 IC50: 5.2 nM (Wee1) Purity:98% MK-1775 is a potent Wee1 inhibitor with IC 50 of 5.2 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2669 Supplier Medchemexpress LLC Supplier No. HY10993100MG
Medchemexpress, HY-10993 100mg MK-1775 AZD1775;Adavosertib CAS:955365-80-7 Formula:C27H32N8O2 IC50: 5.2 nM (Wee1) Purity:98% MK-1775 is a potent Wee1 inhibitor with IC 50 of 5.2 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.