Learn More
Selleck Chemical LLC Adavosertib (MK-1775) 5mg 955365-80-7 AZD1775
SDP
Catalog No. 508890
Encompass_Preferred
Adavosertib (MK-1775, AZD1775) is a potent and selective Wee1 inhibitor with IC50 of 5.2 nM in a cell-free assay; hinders G2 DNA damage checkpoint. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-889-0 Supplier Selleck Chemical LLC Supplier No. S15255MG
Adavosertib (MK-1775, AZD1775) is a potent and selective Wee1 inhibitor with IC50 of 5.2 nM in a cell-free assay; hinders G2 DNA damage checkpoint. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.