Learn More
Abcam MK 212 hydrochloride, 5-HT2C receptor agonist, 10MG
Catalog No. 502704609
Encompass_Preferred
MW 235.11 Da, Purity >99%. Potent, selective 5-HT2C receptor agonist (IC50 values are 28 and 420 nM for 5-HT2C and 5-HT2A receptors, respectively). Shows anxiogenic effects and decreases locomotor activity *in vivo.*.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4609 Supplier Abcam Supplier No. AB14643910MG
MW 235.11 Da, Purity >99%. Potent, selective 5-HT2C receptor agonist (IC50 values are 28 and 420 nM for 5-HT2C and 5-HT2A receptors, respectively). Shows anxiogenic effects and decreases locomotor activity *in vivo.*.
The product is subject to the following: Abcam Restricted Use Statement