Learn More
Enzo Life Sciences MK-2206 2HCl (5mg). CAS: 1032350-13-2
SDP
Catalog No. 502013471
Encompass_Preferred
MK-2206 is a highly selective inhibitor of Akt1, Akt2 and Akt3 with IC50 of 8 nM, 12 nM and 65 nM, respectively. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO (14mg/ml) or ethanol (1mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-3471 Supplier Enzo Life Sciences Supplier No. ENZCHM1640005
MK-2206 is a highly selective inhibitor of Akt1, Akt2 and Akt3 with IC50 of 8 nM, 12 nM and 65 nM, respectively. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO (14mg/ml) or ethanol (1mg/ml). Long Term Storage: -20°C.