Learn More
Selleck Chemical LLC MK-2206 2HCl 5mg 1032350-13-2
SDP
Catalog No. 508725
Encompass_Preferred
MK-2206 2HCl is a highly selective inhibitor of Akt1/2/3 with IC50 of 8 nM/12 nM/65 nM in cell-free assays, respectively; no inhibitory activities against 250 other protein kinases observed. MK-2206 2HCl induces autophagy and apoptosis in cancer cells. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-872-5 Supplier Selleck Chemical LLC Supplier No. S10785MG
MK-2206 2HCl is a highly selective inhibitor of Akt1/2/3 with IC50 of 8 nM/12 nM/65 nM in cell-free assays, respectively; no inhibitory activities against 250 other protein kinases observed. MK-2206 2HCl induces autophagy and apoptosis in cancer cells. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.