Learn More
Medchemexpress LLC HY-10358 200mg , MK-2206 (dihydrochloride) CAS:1032350-13-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10358200MG
Encompass_Preferred
HY-10358 200mg MK-2206 (dihydrochloride) CAS: 1032350-13-2 MK-2206 dihydrochloride (MK-2206 (2HCl)) is an orally active allosteric AKT inhibitor with IC50s of 5 nM, 12 nM, and 65 nM for AKT1, AKT2, and AKT3, respectively. MK-2206 dihydrochloride induces autophagy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9120