Learn More
Medchemexpress LLC HY-10358 10mg Medchemexpress, MK-2206 (dihydrochloride) CAS:1032350-13-2 Purity:>98%
SDP
Catalog No. 502028732
Encompass_Preferred
Medchemexpress, HY-10358 10mg MK-2206 (dihydrochloride) CAS:1032350-13-2 MK-2206 dihydrochloride (MK-2206 (2HCl)) is an orally active allosteric AKT inhibitor with IC50s of 5 nM, 12 nM, and 65 nM for AKT1, AKT2, and AKT3, respectively. MK-2206 dihydrochloride induces autophagy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8732 Supplier Medchemexpress LLC Supplier No. HY1035810MG
Medchemexpress, HY-10358 10mg MK-2206 (dihydrochloride) CAS:1032350-13-2 MK-2206 dihydrochloride (MK-2206 (2HCl)) is an orally active allosteric AKT inhibitor with IC50s of 5 nM, 12 nM, and 65 nM for AKT1, AKT2, and AKT3, respectively. MK-2206 dihydrochloride induces autophagy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.