Learn More
TARGETMOL CHEMICALS INC MK-2206 DIHYDROCHLORIDE 50MG
SDP
Catalog No. 502581047
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MK-2206 dihydrochloride (MK-2206 2HCl) is a highly specific inhibitor of Akt1/2/3 (IC50 8/12/65 nM in cell-free assays) and no effect on the activities of 250 other protein kinases. purity: 99%
Catalog No. 50-258-1047 Supplier TARGETMOL CHEMICALS INC Supplier No. T195250MG
Also available in 2 mg 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MK-2206 dihydrochloride (MK-2206 2HCl) is a highly specific inhibitor of Akt1/2/3 (IC50 8/12/65 nM in cell-free assays) and no effect on the activities of 250 other protein kinases. purity: 99%