Learn More
BIOGEMS INTERNATIONAL INC MK-2206 dihydrochloride 5mg
SDP
Catalog No. 501529559
Encompass
MK-2206 is a highly allosteric selective Akt1,2,3 (IC50 5nM, 12nM, 65nM) inhibitor. It is reported to inhibit the proliferation of cancer cell lines when used synergistically with cytotoxic agents such as erlotinib or lapatinib and significantly enhances cell apoptosis.
Catalog No. 50-152-9559 Supplier BIOGEMS INTERNATIONAL INC Supplier No. 10313205MG
MK-2206 is a highly allosteric selective Akt1,2,3 (IC50 5nM, 12nM, 65nM) inhibitor. It is reported to inhibit the proliferation of cancer cell lines when used synergistically with cytotoxic agents such as erlotinib or lapatinib and significantly enhances cell apoptosis.