Learn More
Medchemexpress LLC HY-10358 100mg , MK 2206 (dihydrochloride) CAS:1032350-13-2 Purity:98%
SDP
Catalog No. 501492671
Encompass_Preferred
Medchemexpress, HY-10358 100mg MK 2206 (dihydrochloride) CAS:1032350-13-2 Formula:C25H23Cl2N5O IC50: 5 nM/12 nM/65 nM (Akt1/Akt2/Akt3) Purity:98% MK 2206 is an orally active allosteric Akt inhibitor with IC 50 of 5 nM/12 nM/65 nM for Akt1/Akt2/Akt3 , respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2671 Supplier Medchemexpress LLC Supplier No. HY10358100MG
Medchemexpress, HY-10358 100mg MK 2206 (dihydrochloride) CAS:1032350-13-2 Formula:C25H23Cl2N5O IC50: 5 nM/12 nM/65 nM (Akt1/Akt2/Akt3) Purity:98% MK 2206 is an orally active allosteric Akt inhibitor with IC 50 of 5 nM/12 nM/65 nM for Akt1/Akt2/Akt3 , respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.