Learn More
Cayman Chemical MK-2206 (hydrochloride), 1032350-13-2, 10 mg
SDP
Supplier:  Cayman Chemical 1159310
Encompass_Preferred
An orally active, allosteric Akt inhibitor (IC50s = 5, 12, and 65 nM for Akt1, 2, and 3, respectively); inhibits cell proliferation of human cancer cell lines synergistically in combination with other topoisomerase inhibitors, antimetabolites, antimicrotubule agents, DNA cross-linkers or growth factor inhibitors
Catalog No. NC1419665