Learn More
Cayman Chemical MK-2206 (hydrochloride), 1032350-13-2, 1 mg
SDP
Supplier:  Cayman Chemical 115931
Encompass_Preferred
Molecular Formula C25H21N5O • 2HCl, Purity ≥98%, Formula Weight 480.4; An orally active, allosteric Akt inhibitor (IC50s = 5, 12, and 65 nM for Akt1, 2, and 3, respectively); inhibits cell proliferation of human cancer cell lines synergistically in combination with other topoisomerase inhibitors, antimetabolites, antimicrotubule agents, DNA cross-linkers or growth factor inhibitors.
Catalog No. NC1316862