Learn More
TARGETMOL CHEMICALS INC MK-2461 5MG
SDP
Catalog No. 502579567
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MK-2461 is a novel ATP-competitive multitargeted inhibitor of activated c-Met with a mean IC50 of 2.5 nM. purity: 99%
Catalog No. 50-257-9567 Supplier TARGETMOL CHEMICALS INC Supplier No. T60945MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MK-2461 is a novel ATP-competitive multitargeted inhibitor of activated c-Met with a mean IC50 of 2.5 nM. purity: 99%