Learn More
Medchemexpress LLC HY-137207 5mg Medchemexpress, MK-28 CAS:864388-65-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1372075MG
Encompass_Preferred
Medchemexpress, HY-137207 5mg MK-28 CAS:864388-65-8 MK-28 is a potent and selective PERK activator. MK-28 exhibits remarkable pharmacokinetic properties and high BBB penetration in mice. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1476