Learn More
Medchemexpress LLC MK-28 | 864388-65-8 | >99.8% | 396.44 | 50 MG
SDP
Catalog No. 5000351773
Encompass_Preferred
MK-28 is a potent and selective PERK activator with remarkable pharmacokinetic properties and high BBB penetration in mice. It improves systemic function and survival in R6/2 mice and induces increased levels of eIF2α-P in the mouse brain striatum. It also significantly improves motor and executive functions and delays death onset in R6/2 mice without observed toxicity.
  • Demonstrates PERK selectivity on a 391-kinase panel
  • Rescues cells from ER stress-induced apoptosis
  • Increases ATF4 protein levels significantly, up to 2.5-fold
  • Significantly increases CHOP and GADD34 mRNA levels
  • Minimal effect on EIF2AK1 (HRI) or EIF2AK2 (PKR)
  • Activates EIF2AK4 (GCN2)
  • Improves systemic function and survival in R6/2 mice
  • Induces increased levels of eIF2α-P in the mouse brain striatum
  • Significantly enhances motor and executive functions
  • Delays death onset in R6/2 mice with no observed toxicity
Catalog No. 50-003-51773 Supplier Medchemexpress LLC Supplier No. HY13720750MG
MK-28 is a potent and selective PERK activator with remarkable pharmacokinetic properties and high BBB penetration in mice. It improves systemic function and survival in R6/2 mice and induces increased levels of eIF2α-P in the mouse brain striatum. It also significantly improves motor and executive functions and delays death onset in R6/2 mice without observed toxicity.
  • Demonstrates PERK selectivity on a 391-kinase panel
  • Rescues cells from ER stress-induced apoptosis
  • Increases ATF4 protein levels significantly, up to 2.5-fold
  • Significantly increases CHOP and GADD34 mRNA levels
  • Minimal effect on EIF2AK1 (HRI) or EIF2AK2 (PKR)
  • Activates EIF2AK4 (GCN2)
  • Improves systemic function and survival in R6/2 mice
  • Induces increased levels of eIF2α-P in the mouse brain striatum
  • Significantly enhances motor and executive functions
  • Delays death onset in R6/2 mice with no observed toxicity