Learn More
Medchemexpress LLC Mk-28 | 864388-65-8 | 99.88% | 396.44 | 100 MG
SDP
Catalog No. 5000351515
Encompass_Preferred
MK-28 is a potent and selective PERK activator that demonstrates significant pharmacokinetic properties and high blood-brain barrier (BBB) penetration in mice. It has been shown to rescue cells from ER stress-induced apoptosis in vitro and improve systemic function and survival in R6/2 mice, an animal model for Huntington's disease. MK-28 also enhances motor and executive functions and delays the onset of death in R6/2 mice without causing toxicity.
  • Potent and selective PERK activator
  • Exhibits significant pharmacokinetic properties
  • High blood-brain barrier (BBB) penetration in mice
  • Rescues cells from ER stress-induced apoptosis in vitro
  • Improves systemic function and survival in R6/2 mice
  • Enhances motor and executive functions and delays death onset in R6/2 mice
  • Activates EIF2AK4 (GCN2)
  • Has little or no effect on EIF2AK1 (HRI) or EIF2AK2 (PKR)
Catalog No. 50-003-51515 Supplier Medchemexpress LLC Supplier No. HY137207100MG
MK-28 is a potent and selective PERK activator that demonstrates significant pharmacokinetic properties and high blood-brain barrier (BBB) penetration in mice. It has been shown to rescue cells from ER stress-induced apoptosis in vitro and improve systemic function and survival in R6/2 mice, an animal model for Huntington's disease. MK-28 also enhances motor and executive functions and delays the onset of death in R6/2 mice without causing toxicity.
  • Potent and selective PERK activator
  • Exhibits significant pharmacokinetic properties
  • High blood-brain barrier (BBB) penetration in mice
  • Rescues cells from ER stress-induced apoptosis in vitro
  • Improves systemic function and survival in R6/2 mice
  • Enhances motor and executive functions and delays death onset in R6/2 mice
  • Activates EIF2AK4 (GCN2)
  • Has little or no effect on EIF2AK1 (HRI) or EIF2AK2 (PKR)