Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MK-3207 hydrochloride | 957116-20-0 | MFCD16038934 | 98.1% | 594.05 g/mol | C31H30ClF2N5O3 | 1 ML
SDP

Catalog No. 5000246271
Encompass_Preferred

MK-3207 Hydrochloride is a potent, orally bioavailable CGRP receptor antagonist used in research on CGRP-mediated signaling and migraine-related pathways. It exhibits sub-nanomolar potency and high selectivity versus related receptors, and is provided in solution and solid forms for laboratory use.

  • Potent CGRP receptor antagonist with IC50 0.12 nM and Ki 0.024 nM.
  • Highly selective versus AM1, AM2, CTR, and AMY3 receptors.
  • Available as a 10 mM solution (1 mL) and as solid quantities for preparation.
  • Purity approximately 98.1% by analytical methods.
  • Molecular formula C31H30ClF2N5O3; molecular weight 594.05 g/mol.
  • Provided with structural identifiers to support research (SMILES, PubChem CID).

Catalog No. 50-002-46271 Supplier Medchemexpress LLC Supplier No. HY1030210MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MK-3207 Hydrochloride is a potent, orally bioavailable CGRP receptor antagonist used in research on CGRP-mediated signaling and migraine-related pathways. It exhibits sub-nanomolar potency and high selectivity versus related receptors, and is provided in solution and solid forms for laboratory use.

  • Potent CGRP receptor antagonist with IC50 0.12 nM and Ki 0.024 nM.
  • Highly selective versus AM1, AM2, CTR, and AMY3 receptors.
  • Available as a 10 mM solution (1 mL) and as solid quantities for preparation.
  • Purity approximately 98.1% by analytical methods.
  • Molecular formula C31H30ClF2N5O3; molecular weight 594.05 g/mol.
  • Provided with structural identifiers to support research (SMILES, PubChem CID).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.