Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MK-3207 hydrochloride | 957116-20-0 | MFCD16038934 | 98.1% | 594.05 g/mol | C31H30ClF2N5O3 | 5 MG
SDP

Catalog No. 5000246142
Encompass_Preferred

MK-3207 hydrochloride is a potent, orally bioavailable antagonist of the calcitonin gene-related peptide (CGRP) receptor used in preclinical research to investigate CGRP-mediated signaling, migraine models, and receptor pharmacology.

  • High affinity CGRP receptor antagonist (IC50 0.12 nM; Ki 0.024 nM).
  • Purity 98.1%.
  • Molecular formula C31H30ClF2N5O3; molecular weight 594.05 g/mol.
  • Registered CAS number 957116-20-0.
  • Suitable for in vitro and in vivo preclinical studies.

Catalog No. 50-002-46142 Supplier Medchemexpress LLC Supplier No. HY103025MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MK-3207 hydrochloride is a potent, orally bioavailable antagonist of the calcitonin gene-related peptide (CGRP) receptor used in preclinical research to investigate CGRP-mediated signaling, migraine models, and receptor pharmacology.

  • High affinity CGRP receptor antagonist (IC50 0.12 nM; Ki 0.024 nM).
  • Purity 98.1%.
  • Molecular formula C31H30ClF2N5O3; molecular weight 594.05 g/mol.
  • Registered CAS number 957116-20-0.
  • Suitable for in vitro and in vivo preclinical studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.