Learn More
Selleck Chemical LLC MK-3903 5mg 1219737-12-8
SDP
Supplier:  Selleck Chemical LLC S88035MG
Encompass_Preferred
MK-3903 is a potent and selective AMPK activator with an EC50 of 8 nM for ?1 ?1 ?1 subunit. It activates 10 of the 12 pAMPK complexes with EC50 values in the range of 8-40 nM and maximal activation >50%. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8377