Learn More
Medchemexpress LLC HY-107709 100mg Medchemexpress, MK-4074 CAS:1039758-22-9 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY107709100MG
Encompass_Preferred
Medchemexpress,HY-107709 100mg MK-4074 CAS:1039758-22-9 Formula:C33H31N3O6 MK-4074 is a liver-specific inhibitor of acetyl-CoA carboxylase ACC1 and ACC2 with IC50 values of approximately 3 nM. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-7357