Learn More
Medchemexpress LLC MK-4166 5mg | 5mg
SDP
Catalog No. 5000323328
Encompass_Preferred
MK-4166 is a humanized IgG1 agonist monoclonal antibody targeting GITR MK-4166 enhances the proliferation of both na ve and tumor-infiltrating T lymphocytes[1]
Catalog No. 50-003-23328 Supplier Medchemexpress LLC Supplier No. HYP9911795MG
MK-4166 is a humanized IgG1 agonist monoclonal antibody targeting GITR MK-4166 enhances the proliferation of both na ve and tumor-infiltrating T lymphocytes[1]