Learn More
Medchemexpress LLC HY-13466 1mg Medchemexpress, MK-4256 CAS:1104599-69-0 Purity:>98%
SDP
Catalog No. 502081373
Encompass_Preferred
Medchemexpress, HY-13466 1mg MK-4256 CAS:1104599-69-0 MK-4256 is a potent and selective SSTR3 antagonist with IC50s of 0.66 nM and 0.36 nM in human and mouse receptor binding assays, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1373 Supplier Medchemexpress LLC Supplier No. HY134661MG
Medchemexpress, HY-13466 1mg MK-4256 CAS:1104599-69-0 MK-4256 is a potent and selective SSTR3 antagonist with IC50s of 0.66 nM and 0.36 nM in human and mouse receptor binding assays, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.