Learn More
Medchemexpress LLC HY-10619 100mg Medchemexpress, MK-4827 Niraparib CAS:1038915-60-4 Purity:98%
SDP
Catalog No. 501637305
Encompass_Preferred
Medchemexpress,HY-10619 100mg MK-4827 Niraparib CAS:1038915-60-4 Formula:C19H20N4O MK-4827 (Niraparib) is a highly potent PARP1 and PARP2 inhibitor with IC50s of 3.8 and 2.1 nM, respectively. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-7305 Supplier Medchemexpress LLC Supplier No. HY10619100MG
Medchemexpress,HY-10619 100mg MK-4827 Niraparib CAS:1038915-60-4 Formula:C19H20N4O MK-4827 (Niraparib) is a highly potent PARP1 and PARP2 inhibitor with IC50s of 3.8 and 2.1 nM, respectively. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.