Learn More
Cayman Chemical MK-4827tosylate 10mg
SDP
Catalog No. 502982273
Encompass_Preferred
An orally bioavailable inhibitor of PARP1 and PARP2 (IC50s = 3.8 and 2.1 nM, respectively); inhibits the proliferation of BRCA1 and BRCA2 mutant cancer cells in vitro (IC50s = 10-100 nM) and has efficacy alone against a xenograft of BRCA1-deficient cancer in mice
Catalog No. 50-298-2273 Supplier Cayman Chemical Supplier No. 2084210 MG
An orally bioavailable inhibitor of PARP1 and PARP2 (IC50s = 3.8 and 2.1 nM, respectively); inhibits the proliferation of BRCA1 and BRCA2 mutant cancer cells in vitro (IC50s = 10-100 nM) and has efficacy alone against a xenograft of BRCA1-deficient cancer in mice