Learn More
TARGETMOL CHEMICALS INC MK-5108 10MG
SDP
Catalog No. 502576610
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MK-5108 (VX-689) is a highly potent and specific Aurora-A kinase inhibitor with an IC50 value of 0.064 nM. purity: 100%
Catalog No. 50-257-6610 Supplier TARGETMOL CHEMICALS INC Supplier No. T606810MG
Add to Cart
Add to Cart
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MK-5108 (VX-689) is a highly potent and specific Aurora-A kinase inhibitor with an IC50 value of 0.064 nM. purity: 100%