Learn More
Medchemexpress LLC HY-13252 10mg Medchemexpress, MK-5108 CAS:1010085-13-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1325210MG
Encompass_Preferred
Medchemexpress, HY-13252 10mg MK-5108 CAS:1010085-13-8 MK-5108 is a highly potent and specific inhibitor of Aurora A kinase with an IC50 value of 0.064 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8971