Learn More
Medchemexpress LLC HY-13252 5mg , MK-5108 CAS:1010085-13-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY132525MG
Encompass_Preferred
HY-13252 5mg MK-5108 CAS: 1010085-13-8 MK-5108 is a highly potent and specific inhibitor of Aurora A kinase with an IC50 value of 0.064 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9691