Learn More
Selleck Chemical LLC MK-5108 S2770-1g
SDP
Catalog No. 503111806
Encompass_Preferred
MK-5108 is a highly selective Aurora A inhibitor with IC50 of 0 064 nM in a cell-free assay and is 220- and 190-fold more selective for Aurora A than Aurora B/C while it inhibits TrkA with less than 100-fold selectivity MK-5108 (VX-689) induces autophagy Phase 1
Catalog No. 50-311-1806 Supplier Selleck Chemical LLC Supplier No. S27701G
Add to Cart
Add to Cart
MK-5108 is a highly selective Aurora A inhibitor with IC50 of 0 064 nM in a cell-free assay and is 220- and 190-fold more selective for Aurora A than Aurora B/C while it inhibits TrkA with less than 100-fold selectivity MK-5108 (VX-689) induces autophagy Phase 1