Learn More
Apexbio Technology LLC MK-5108 (VX-689) 1010085-13-8 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061541
Encompass_Preferred
MK-5108 (VX-689 CAS 1010085-13-8) is a selective inhibitor targeting Aurora A kinase (AAK) functioning through competitive interaction with its ATP-binding pocket It exhibits sub-nanomolar inhibition of Aurora A kinase (IC50 0 064 nM) and displays additional inhibitory activity against Aurora B and Aurora C kinases (IC50 14 nM and IC50 12 nM respectively) MK-5108 has been extensively studied in cancer research contexts demonstrating antiproliferative activity across various tumor types including breast cervical colorectal ovarian and pancreatic cancers
Catalog No. 50-000-61541 Supplier Apexbio Technology LLC Supplier No. A41205.1
MK-5108 (VX-689 CAS 1010085-13-8) is a selective inhibitor targeting Aurora A kinase (AAK) functioning through competitive interaction with its ATP-binding pocket It exhibits sub-nanomolar inhibition of Aurora A kinase (IC50 0 064 nM) and displays additional inhibitory activity against Aurora B and Aurora C kinases (IC50 14 nM and IC50 12 nM respectively) MK-5108 has been extensively studied in cancer research contexts demonstrating antiproliferative activity across various tumor types including breast cervical colorectal ovarian and pancreatic cancers