Learn More
Selleck Chemical LLC MK-5108 (VX-689) 5mg 1010085-13-8
SDP
Supplier:  Selleck Chemical LLC S27705MG
Encompass_Preferred
MK-5108 (VX-689) is a highly selective Aurora A inhibitor with IC50 of 0.064 nM in a cell-free assay and is 220- and 190-fold more selective for Aurora A than Aurora B/C, while it inhibits TrkA with less than 100-fold selectivity. MK-5108 (VX-689) induces autophagy. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0446835