Learn More
Apexbio Technology LLC MK-5172 1350514-68-9 10mM (in 1mL DMSO)
SDP
Catalog No. 5000064284
Encompass_Preferred
MK-5172 is a selective inhibitor targeting the hepatitis C virus (HCV) NS3/4A protease an enzyme essential for viral replication The HCV virus genome encodes this protease which cleaves polyprotein precursors enabling functional viral proteins formation during replication MK-5172 inhibits protease activity across multiple HCV genotypes demonstrating IC50 values in enzymatic assays at low nanomolar concentrations In cell-based replicon systems MK-5172 exhibits IC50 values of 2 nM (genotype 1a) 0 5 nM (genotype 1b) 8 nM (genotype 2a) and 13 nM (genotype 3) MK-5172 is utilized in antiviral studies examining various genotype infections viral resistant variants and the dynamics of viral load reductions in experimental chronic infections
Catalog No. 50-000-64284 Supplier Apexbio Technology LLC Supplier No. A36185.1
MK-5172 is a selective inhibitor targeting the hepatitis C virus (HCV) NS3/4A protease an enzyme essential for viral replication The HCV virus genome encodes this protease which cleaves polyprotein precursors enabling functional viral proteins formation during replication MK-5172 inhibits protease activity across multiple HCV genotypes demonstrating IC50 values in enzymatic assays at low nanomolar concentrations In cell-based replicon systems MK-5172 exhibits IC50 values of 2 nM (genotype 1a) 0 5 nM (genotype 1b) 8 nM (genotype 2a) and 13 nM (genotype 3) MK-5172 is utilized in antiviral studies examining various genotype infections viral resistant variants and the dynamics of viral load reductions in experimental chronic infections