Learn More
Apexbio Technology LLC MK-5172, 5mg. Cas: 1350514-68-9 MFCD: MFCD25976700. HCV NS3/4a protease inhibitor
SDP
Supplier:  Apexbio Technology LLC A36185
Encompass_Preferred
MK-5172 is a selective inhibitor of Hepatitis C Virus NS3/4a Protease [1].Hepatitis C (HCV) virus is a member of the Flaviviridae family of viruses in the Hepacivirus genus and encoded by a 9.6-kb positive strand RNA genome [2]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4818