Learn More
Medchemexpress LLC Mk-571 (l-660711) | 115104-28-4 | >98.9% | 515.09 | 25 MG
SDP
Catalog No. 5000402734
Encompass_Preferred
MK-571 (L-660711) is an orally active, potent, and selective competitive leukotriene D4 (LTD4) receptor antagonist. It also acts as an MRP4 and ABCC1 (MRP1) inhibitor and inhibits constitutive and antigen-stimulated S1P (sphingosine-1-phosphate) release.
  • Orally active.
  • Potent and selective competitive leukotriene D4 (LTD4) receptor antagonist.
  • Inhibitor of MRP4 and ABCC1 (MRP1).
  • Inhibits constitutive and antigen-stimulated S1P (sphingosine-1-phosphate) release.
  • Potential for inhibiting antigen-induced dyspnea.
  • Potential for blocking LTD4- and Ascaris-induced bronchoconstriction.
  • Potential for reversal of hypoxic pulmonary hypertension (PH) and protection from hypoxic PH.
Catalog No. 50-004-02734 Supplier Medchemexpress LLC Supplier No. 25MGHY19989
MK-571 (L-660711) is an orally active, potent, and selective competitive leukotriene D4 (LTD4) receptor antagonist. It also acts as an MRP4 and ABCC1 (MRP1) inhibitor and inhibits constitutive and antigen-stimulated S1P (sphingosine-1-phosphate) release.
  • Orally active.
  • Potent and selective competitive leukotriene D4 (LTD4) receptor antagonist.
  • Inhibitor of MRP4 and ABCC1 (MRP1).
  • Inhibits constitutive and antigen-stimulated S1P (sphingosine-1-phosphate) release.
  • Potential for inhibiting antigen-induced dyspnea.
  • Potential for blocking LTD4- and Ascaris-induced bronchoconstriction.
  • Potential for reversal of hypoxic pulmonary hypertension (PH) and protection from hypoxic PH.