Learn More
Medchemexpress LLC Mk-571 (L-660711) | 115104-28-4 | MFCD00797675 | 98.9% | 515.09 g/mol | C26H27ClN2O3S2 | 10 MG
SDP
Catalog No. 5000297595
Encompass_Preferred
MK-571 is an orally active, potent, and selective competitive leukotriene D4 (LTD4) receptor antagonist that also inhibits ABCC1/MRP4 transporters. Supplied for research use only, it is used in pharmacology and cell biology studies to probe leukotriene signaling and transporter-mediated processes.
  • Potent and selective leukotriene D4 receptor antagonist.
  • Inhibits ABCC1/MRP4 transporters.
  • High purity (≈98.9%).
  • Solid, off-white to light yellow appearance.
  • Stable under recommended storage; protect from light.
Catalog No. 50-002-97595 Supplier Medchemexpress LLC Supplier No. HY1998910MG
MK-571 is an orally active, potent, and selective competitive leukotriene D4 (LTD4) receptor antagonist that also inhibits ABCC1/MRP4 transporters. Supplied for research use only, it is used in pharmacology and cell biology studies to probe leukotriene signaling and transporter-mediated processes.
  • Potent and selective leukotriene D4 receptor antagonist.
  • Inhibits ABCC1/MRP4 transporters.
  • High purity (≈98.9%).
  • Solid, off-white to light yellow appearance.
  • Stable under recommended storage; protect from light.