Learn More
Medchemexpress LLC MK-571 (sodium) | 115103-85-0 | 99.8% | 537.07 | 1 ML
SDP
Catalog No. 5000404554
Encompass_Preferred
MK-571 sodium is an orally active, potent, and selective competitive leukotriene D4 (LTD4) receptor antagonist. It also acts as an inhibitor of multidrug resistance-associated protein MRP4 (ABCC4) and ABCC1 (MRP1), and inhibits constitutive and antigen-stimulated sphingosine-1-phosphate (S1P) release.
  • Potent and selective leukotriene D4 (LTD4) receptor antagonist.
  • Inhibitor of multidrug resistance-associated protein MRP4 (ABCC4).
  • Inhibitor of ABCC1 (MRP1).
  • Inhibits constitutive and antigen-stimulated sphingosine-1-phosphate (S1P) release.
  • Demonstrates activity in in vitro and in vivo studies.
Catalog No. 50-004-04554 Supplier Medchemexpress LLC Supplier No. HY19989A10MM/1ML
MK-571 sodium is an orally active, potent, and selective competitive leukotriene D4 (LTD4) receptor antagonist. It also acts as an inhibitor of multidrug resistance-associated protein MRP4 (ABCC4) and ABCC1 (MRP1), and inhibits constitutive and antigen-stimulated sphingosine-1-phosphate (S1P) release.
  • Potent and selective leukotriene D4 (LTD4) receptor antagonist.
  • Inhibitor of multidrug resistance-associated protein MRP4 (ABCC4).
  • Inhibitor of ABCC1 (MRP1).
  • Inhibits constitutive and antigen-stimulated sphingosine-1-phosphate (S1P) release.
  • Demonstrates activity in in vitro and in vivo studies.