Learn More
Medchemexpress LLC MK-571 sodium | 115103-85-0 | 99.8% | 537.07 | 5 MG
SDP
Catalog No. 5000401110
Encompass_Preferred
MK-571 (L-660711) sodium is an orally active, potent, and selective competitive leukotriene D4 (LTD4) receptor antagonist. It inhibits constitutive and antigen-stimulated S1P (sphingosine-1-phosphate) release.
  • Orally active, potent and selective competitive leukotriene D4 (LTD4) receptor antagonist.
  • Ki values of 0.22 nM in guinea pig lung membranes and 2.1 nM in human lung membranes for LTD4.
  • Inhibitor of multidrug resistance-associated protein MRP4 (ABCC4) and ABCC1 (MRP1).
  • Inhibits constitutive and antigen-stimulated S1P (sphingosine-1-phosphate) release.
Catalog No. 50-004-01110 Supplier Medchemexpress LLC Supplier No. HY19989A5MG
Add to Cart
Add to Cart
MK-571 (L-660711) sodium is an orally active, potent, and selective competitive leukotriene D4 (LTD4) receptor antagonist. It inhibits constitutive and antigen-stimulated S1P (sphingosine-1-phosphate) release.
  • Orally active, potent and selective competitive leukotriene D4 (LTD4) receptor antagonist.
  • Ki values of 0.22 nM in guinea pig lung membranes and 2.1 nM in human lung membranes for LTD4.
  • Inhibitor of multidrug resistance-associated protein MRP4 (ABCC4) and ABCC1 (MRP1).
  • Inhibits constitutive and antigen-stimulated S1P (sphingosine-1-phosphate) release.