Learn More
Medchemexpress LLC MK-571 (sodium) | 115103-85-0 | L-660711 (sodium) | 99.75% | 537.07 | 50 MG
SDP
Catalog No. 5000405086
Encompass_Preferred
MK-571 (L-660711) sodium is an orally active, potent, and selective competitive leukotriene D4 (LTD4) receptor antagonist, with Ki values of 0.22 and 2.1 nM in guinea pig and human lung membranes, respectively. It is also an inhibitor of multidrug resistance-associated protein MRP4 (ABCC4) and ABCC1 (MRP1). MK-571 sodium inhibits constitutive and antigen-stimulated S1P (sphingosine-1-phosphate) release.
  • Orally active.
  • Potent and selective competitive leukotriene D4 (LTD4) receptor antagonist.
  • Inhibitor of multidrug resistance-associated protein MRP4 (ABCC4) and ABCC1 (MRP1).
  • Inhibits constitutive and antigen-stimulated S1P (sphingosine-1-phosphate) release.
  • Appearance: solid, light yellow to yellow.
Catalog No. 50-004-05086 Supplier Medchemexpress LLC Supplier No. HY19989A50MG
MK-571 (L-660711) sodium is an orally active, potent, and selective competitive leukotriene D4 (LTD4) receptor antagonist, with Ki values of 0.22 and 2.1 nM in guinea pig and human lung membranes, respectively. It is also an inhibitor of multidrug resistance-associated protein MRP4 (ABCC4) and ABCC1 (MRP1). MK-571 sodium inhibits constitutive and antigen-stimulated S1P (sphingosine-1-phosphate) release.
  • Orally active.
  • Potent and selective competitive leukotriene D4 (LTD4) receptor antagonist.
  • Inhibitor of multidrug resistance-associated protein MRP4 (ABCC4) and ABCC1 (MRP1).
  • Inhibits constitutive and antigen-stimulated S1P (sphingosine-1-phosphate) release.
  • Appearance: solid, light yellow to yellow.