Learn More
Medchemexpress LLC MK-571 sodium | 115103-85-0 | MFCD00878420 | 99.8% | 537.07 | C26H26ClN2NaO3S2 | 10 MG
SDP
Catalog No. 5000297586
Encompass_Preferred
MK-571 sodium is a research-grade small molecule inhibitor that functions as a potent leukotriene D4 (LTD4) receptor antagonist and an inhibitor of multidrug resistance-associated proteins. Supplied as a high-purity solid for in vitro and in vivo studies, it is commonly used to probe leukotriene signaling and transporter-mediated processes in pharmacology research.
  • Potent leukotriene D4 receptor antagonist with low nanomolar affinity.
  • Inhibits multidrug resistance-associated proteins (MRP4/ABCC4 and ABCC1).
  • High purity solid suitable for analytical and biological assays.
  • Soluble in DMSO and formulatable for in vivo dosing.
  • Stable under recommended storage conditions for research use.
Catalog No. 50-002-97586 Supplier Medchemexpress LLC Supplier No. HY19989A10MG
MK-571 sodium is a research-grade small molecule inhibitor that functions as a potent leukotriene D4 (LTD4) receptor antagonist and an inhibitor of multidrug resistance-associated proteins. Supplied as a high-purity solid for in vitro and in vivo studies, it is commonly used to probe leukotriene signaling and transporter-mediated processes in pharmacology research.
  • Potent leukotriene D4 receptor antagonist with low nanomolar affinity.
  • Inhibits multidrug resistance-associated proteins (MRP4/ABCC4 and ABCC1).
  • High purity solid suitable for analytical and biological assays.
  • Soluble in DMSO and formulatable for in vivo dosing.
  • Stable under recommended storage conditions for research use.