Learn More
Medchemexpress LLC MK-571 sodium | 115103-85-0 | MFCD11114392 | 99.8% | 537.07 | 100 MG
SDP
Catalog No. 5000402046
Encompass_Preferred
MK-571 sodium (L-660711 sodium) is a potent and selective competitive leukotriene D4 (LTD4) receptor antagonist. It also acts as an inhibitor of multidrug resistance-associated protein MRP4 (ABCC4) and ABCC1 (MRP1). Furthermore, it inhibits constitutive and antigen-stimulated sphingosine-1-phosphate (S1P) release.
  • Orally active.
  • Potent and selective competitive leukotriene D4 (LTD4) receptor antagonist.
  • Inhibits multidrug resistance-associated protein MRP4 (ABCC4) and ABCC1 (MRP1).
  • Inhibits constitutive and antigen-stimulated sphingosine-1-phosphate (S1P) release.
Catalog No. 50-004-02046 Supplier Medchemexpress LLC Supplier No. HY19989A100MG
MK-571 sodium (L-660711 sodium) is a potent and selective competitive leukotriene D4 (LTD4) receptor antagonist. It also acts as an inhibitor of multidrug resistance-associated protein MRP4 (ABCC4) and ABCC1 (MRP1). Furthermore, it inhibits constitutive and antigen-stimulated sphingosine-1-phosphate (S1P) release.
  • Orally active.
  • Potent and selective competitive leukotriene D4 (LTD4) receptor antagonist.
  • Inhibits multidrug resistance-associated protein MRP4 (ABCC4) and ABCC1 (MRP1).
  • Inhibits constitutive and antigen-stimulated sphingosine-1-phosphate (S1P) release.